Learn More
Invitrogen™ D4-GDI Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527873
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji , mouse fibroblast. Predicted reactivity: Mouse (90%), Rat (89%), Xenopus laevis (81%), Pig (94%), Chicken (92%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
Specifications
| D4-GDI | |
| Polyclonal | |
| Unconjugated | |
| ARHGDIB | |
| ARHGDIB; D4; d4 gdi; GDI LYGDI; GDIA2; GDID4; LYGDI; Ly-GDI; RAP1GN1; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP dissociation inhibitor beta; rho GDP-dissociation inhibitor 2; Rho, GDP dissociation inhibitor (GDI) beta; rho-GDI beta; RhoGDI2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11857, 397 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P52566, Q61599 | |
| ARHGDIB | |
| Recombinant protein encompassing a sequence within the center region of human Rho GDI2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.