Learn More
3M™ Secure Click™ Particulate Filter with Nuisance Level Organic Vapour/Acid Gas Relief
Click to view available options
Filter Type:
P2R Nuis OV/AG
P3R Nuis OV/AG
Unit Size:
10 pairs
Description
- New dual flow cartridges and filters allow for easier breathing
- Protects against particulates with optional nuisance level organic vapour/acid gas relief
- Quick, simple connection. Simply align the connections and push until you hear a click
- Use with 3M™ Secure Click™ Reusable Respirators 800 Series
- Swept-back design allows an enhanced field of view and comfort
Specifications
Specifications
| Certifications/Compliance | CE Approved |
| Connection Type | Secure Click™ |
| Filter Type | P2R Nuis OV/AG |
| Product Type | Filter |
| Model | D3128 |
| Quantity | 80/Cs. |