Learn More
Fisherbrand™ D3-14, A3-1 and A6 Wide Gel Electrophoresis System Wall Comb, 1 Tooth, 1.5mm thick tooth
Micro Well and Wall Combs for use with the Fisherbrand™ D3-14, A3-1 and A6 Wide Gel Electrophoresis Systems.
Brand: Fisherbrand™
Code : 04
Additional Details : Weight : 0.01000kg
Description
Micro Well and Wall Combs for use with the Fisherbrand™ D3-14, A3-1 and A6 Wide Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Cat. Nos. D3-MTC, -MTD, -MT2C and -MT2D are 8 and 12-channel pipette format (1X) and (2X) combs
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
Specifications
Wall comb | |
Comb | |
1.5 mm |
Owl™ D3-14, A3-1 and A6 Wide Gel Electrophoresis System | |
1 |