missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ D2HGDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545860
This item is not returnable.
View return policy
Description
Peptide sequence: QESPFYVLIE TSGSNAGHDA EKLGHFLEHA LGSGLVTDGT MATDQRKVKM Sequence homology: Dog: 79%; Horse: 86%; Human: 100%; Mouse: 100%; Pig: 77%; Rabbit: 100%; Rat: 92%.
This gene encodes D-2hydroxyglutarate dehydrogenase, a mitochondrial enzyme belonging to the FAD-binding oxidoreductase/transferase type 4 family. This enzyme, which is most active in liver and kidney but also active in heart and brain, converts D-2-hydroxyglutarate to 2-ketoglutarate. Mutations in this gene are present in D-2-hydroxyglutaric aciduria, a rare recessive neurometabolic disorder causing developmental delay, epilepsy, hypotonia, and dysmorphic features.
Specifications
| D2HGDH | |
| Polyclonal | |
| Unconjugated | |
| D2HGDH | |
| AA408776; AA408778; AI325464; D2HGD; D2HGDH; D2hgdh {ECO:0000250; D-2-hydroxyglutarate dehydrogenase; D-2-hydroxyglutarate dehydrogenase, mitochondrial; FLJ42195; LOW QUALITY PROTEIN: D-2-hydroxyglutarate dehydrogenase, mitochondrial; MGC25181; RGD1307976; UniProtKB:Q8N465} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 728294 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N465 | |
| D2HGDH | |
| Synthetic peptide directed towards the C-terminal of human D2HGDH (aa 332-381). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction