missing translation for 'onlineSavingsMsg'
Learn More

D159687, MedChemExpress

Product Code. 16496766
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496766 100 mg 100mg
16479107 5 mg 5mg
16472617 50 mg 50mg
16416528 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16496766 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY15444
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D159687 is a selective PDE4D inhibitor.

D159687 is a selective PDE4D inhibitor.

Specifications

Chemical Name or Material D159687
CAS 1155877-97-6
Color White
Purity Grade Notes Research
Molecular Formula C21H19ClN2O2
Quantity 100 mg
Solubility Information DMSO : 150 mg/mL (408.90 mM; Need ultrasonic)
SMILES NC(NC1=CC=C(CC2=CC(C3=CC(Cl)=CC=C3)=C(OC)C=C2)C=C1)=O
Molecular Weight (g/mol) 366.84
Formula Weight 366.84
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.