missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D14 Rabbit anti-Plant, Polyclonal, Novus Biologicals™
Description
D14 Polyclonal antibody specifically detects D14 in Plant samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | D14 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 50% glycerol, pH7.3 |
| Gene Alias | D88, HTD2, LOC4331983, Protein DWARF 14, Protein DWARF 88, Protein HIGH-TILLERING DWARF 2, Strigolactone esterase D14, strigolactone esterase D14-like |
| Host Species | Rabbit |
| Immunogen | Recombinant protein of Oryza sativa D14. |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?