missing translation for 'onlineSavingsMsg'
Learn More

D14 Rabbit anti-Plant, Polyclonal, Novus Biologicals™

Product Code. 18314516
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18314516 20 μg 20µL
18350893 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18314516 Supplier Novus Biologicals Supplier No. NBP31606120UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

D14 Polyclonal antibody specifically detects D14 in Plant samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen D14
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias D88, HTD2, LOC4331983, Protein DWARF 14, Protein DWARF 88, Protein HIGH-TILLERING DWARF 2, Strigolactone esterase D14, strigolactone esterase D14-like
Host Species Rabbit
Immunogen Recombinant protein of Oryza sativa D14.
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4331983
Target Species Plant
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.