missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Xylose, C.P. grade, 99% max., MP Biomedicals™

Product Code. p-4404582 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g

Product Code. 11407531

Brand: MP Biomedicals 0210330080

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
MDL Number MFCD00064360
Synonym Wood sugar

Specifications

Color White
Assay Percent Range >99%
Quantity 100 g
Formula Weight 150.13
Grade C.P.
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Xylose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.