missing translation for 'onlineSavingsMsg'
Learn More

D-Xylonic Acid Calcium Salt Hydrate, TRC

Product Code. 30453325
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453325 1 g 1g
30450566 2.5 g 2.5g
30451790 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453325 Supplier Toronto Research Chemicals Supplier No. TRCX7500001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72656-08-7
Molecular Formula C10 H18 Ca O12 . 2 H2 O
Molecular Weight (g/mol) 406.35
IUPAC Name bis[[(2R,3S,4R)-2,3,4,5-tetrahydroxypentanoyl]oxy]calcium;dihydrate
SMILES O.O.OC[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)[C@@H](O)[C@H](O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C5H10O6.Ca.2H2O/c2*6-1-2(7)3(8)4(9)5(10)11;;;/h2*2-4,6-9H,1H2,(H,10,11);;2*1H2/q;;+2;;/p-2/t2*2-,3+,4-;;;/m11.../s1
Formula Weight 406.0635
Percent Purity >95%
Chemical Name or Material D-Xylonic Acid, Calcium Salt, Hydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.