missing translation for 'onlineSavingsMsg'
Learn More

D-Valine tert-butyl ester hydrochloride, 95%

Product Code. 15448708
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448708 25 g 25g
15428708 1 g 1g
15438708 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448708 Supplier Thermo Scientific Alfa Aesar Supplier No. H62131.14
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 104944-18-5 | C9H20ClNO2 | 209.71 g/mol

D-Valine tert-butyl ester hydrochloride used as a intermediate in pharmaceutical. Used as a an antitumor drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Valine tert-butyl ester hydrochloride used as a intermediate in pharmaceutical. Used as a an antitumor drug.

Notes
Store at -20°C. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104944-18-5
Molecular Formula C9H20ClNO2
Molecular Weight (g/mol) 209.71
MDL Number MFCD00237308
InChI Key AUIVQIHTTVPKFS-OGFXRTJISA-N
Synonym h-d-val-otbu.hcl, d-valine tert-butyl ester hydrochloride, h-d-val-otbu hcl, r-tert-butyl 2-amino-3-methylbutanoate hydrochloride, tert-butyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, 1,1-dimethylethyl ester, hydrochloride, d-valine tert.butyl ester hydrochloride, d-valine tert.butyl ester hcl, h-d-val-otbucl, h-d-val-otbu. hcl
PubChem CID 45108216
SMILES Cl.CC(C)[C@@H](N)C(=O)OC(C)(C)C

Specifications

IUPAC Name tert-butyl 2-amino-3-methylbutanoate hydrochloride
Quantity 25 g
Formula Weight 209.71
Percent Purity 95%
Chemical Name or Material D-Valine tert-butyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.