missing translation for 'onlineSavingsMsg'
Learn More

D-Valine methyl ester hydrochloride, 99%

Product Code. 11317079
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317079 5 g 5g
11327079 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317079 Supplier Thermo Scientific Alfa Aesar Supplier No. H26883.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7146-15-8 | C6H14ClNO2 | 167.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7146-15-8
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD00237309
InChI Key KUGLDBMQKZTXPW-NUBCRITNSA-N
Synonym d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl
PubChem CID 11984198
SMILES Cl.COC(=O)[C@H](N)C(C)C

Specifications

Melting Point ∽170°C (decomposition)
Beilstein 3912091
IUPAC Name methyl 2-amino-3-methylbutanoate hydrochloridyl
Quantity 5 g
Formula Weight 167.64
Percent Purity 99%
Chemical Name or Material D-Valine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.