missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Valine, 98+%
£23.80 - £208.00
Chemical Identifiers
CAS | 640-68-6 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.15 |
MDL Number | MFCD00064219 |
InChI Key | KZSNJWFQEVHDMF-SCSAIBSYSA-N |
Synonym | d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid |
PubChem CID | 71563 |
ChEBI | CHEBI:27477 |
IUPAC Name | (2R)-2-amino-3-methylbutanoic acid |
SMILES | CC(C)[C@@H](N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453138
|
Thermo Scientific Chemicals
A18894.06 |
5 g |
£23.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463138
|
Thermo Scientific Chemicals
A18894.14 |
25 g |
£71.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473138
|
Thermo Scientific Chemicals
A18894.22 |
100 g |
£208.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
640-68-6 | |
117.15 | |
KZSNJWFQEVHDMF-SCSAIBSYSA-N | |
71563 | |
(2R)-2-amino-3-methylbutanoic acid |
C5H11NO2 | |
MFCD00064219 | |
d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid | |
CHEBI:27477 | |
CC(C)[C@@H](N)C(O)=O |
Specifications
295°C (decomposition) | |
C5H11NO2 | |
1721135 | |
KZSNJWFQEVHDMF-SCSAIBSYSA-N | |
(2R)-2-amino-3-methylbutanoic acid | |
71563 | |
117.15 | |
−27° (c=5 in 5N HCl) |
640-68-6 | |
MFCD00064219 | |
d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acid | |
CC(C)[C@@H](N)C(O)=O | |
117.15 | |
CHEBI:27477 | |
≥98% | |
D-Valine |
Safety and Handling
missing translation for 'einecsNumber' : 211-368-9
missing translation for 'rtecsNumber' : YV9360000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only