missing translation for 'onlineSavingsMsg'
Learn More

D-Tyrosine tert-butyl ester, 98%

Product Code. 15409898
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409898 5 g 5g
15499888 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409898 Supplier Thermo Scientific Alfa Aesar Supplier No. H63274.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 87553-74-0 | C13H19NO3 | 237.30 g/mol

D-Tyrosine tert-butyl ester, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tyrosine tert-butyl ester, is used as a pharmaceutical intermediate.

Solubility
Very slightly soluble in cold water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87553-74-0
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.30
MDL Number MFCD00191035
InChI Key DIGHFXIWRPMGSA-LDGXTIHJNA-N
Synonym h-d-tyr-otbu, tert-butyl 2r-2-amino-3-4-hydroxyphenyl propanoate, d-tyrosine,1,1-dimethylethyl ester, d-tyrosine tert-butyl ester, d-tyrosine tert.butyl ester, d-tyrosine-t-butylester, r-tert-butyl 2-amino-3-4-hydroxyphenyl propanoate, r-2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester
PubChem CID 7408333
IUPAC Name tert-butyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES CC(C)(C)OC(=O)[C@H](N)CC1=CC=C(O)C=C1

Specifications

Solubility Information Very slightly soluble in cold water.
Quantity 5 g
Formula Weight 237.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material D-Tyrosine tert-butyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.