missing translation for 'onlineSavingsMsg'
Learn More

D-Tyrosine methyl ester hydrochloride, 98%

Product Code. 15420429
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420429 5 g 5g
15430429 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420429 Supplier Thermo Scientific Alfa Aesar Supplier No. H63630.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3728-20-9 | C10H14ClNO3 | 231.68 g/mol

D-Tyrosine methyl ester hydrochloride is an intermediate in the synthesis of oxybutynin chloride (O868525), an inhibitor of proliferation and supresses gene expression in bladder smooth muscle cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tyrosine methyl ester hydrochloride is an intermediate in the synthesis of oxybutynin chloride (O868525), an inhibitor of proliferation and supresses gene expression in bladder smooth muscle cells.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3728-20-9
Molecular Formula C10H14ClNO3
Molecular Weight (g/mol) 231.68
MDL Number MFCD00066124
InChI Key VXYFARNRGZWHTJ-MVRPGTNWNA-N
Synonym h-d-tyr-ome.hcl, d-tyrosine methyl ester hydrochloride, h-d-tyr-ome hcl, methyl d-tyrosinate hydrochloride, r-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, d-tyrosine, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-hydroxyphenyl propanoate hydrochloride, methyl d-tyrosinate hcl, tyrosine methylester hydrochloride, h-d-tyr-omehcl
PubChem CID 134565
IUPAC Name methyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)CC1=CC=C(O)C=C1

Specifications

Color White
Quantity 5 g
Formula Weight 231.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Tyrosine methyl ester hydrochloride
compliance-icons
Product Identifier
  • D-Tyrosine methyl ester hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Methyl D-tyrosinatehydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.