Learn More
Thermo Scientific Chemicals D-Turanose, 98%
Description
D-Turanose is used as a carbon source by bacteria, fungi, and other organisms. It is involved in intracellular sugar signaling in plant cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-Turanose is used as a carbon source by bacteria, fungi, and other organisms. It is involved in intracellular sugar signaling in plant cells.
Solubility
Soluble in water and methanol.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container. Store at room temperature.
Specifications
Specifications
| Melting Point | ∼170°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 93771 |
| Merck Index | 14,9821 |
| Solubility Information | Soluble in water and methanol. |
| Formula Weight | 342.3 |
| Percent Purity | 98% |
| Chemical Name or Material | D-Turanose |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.