missing translation for 'onlineSavingsMsg'
Learn More

D-Tryptophan methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15447108
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15447108

Brand: Thermo Scientific Alfa Aesar H60622.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-Tryptophan Methyl Ester Hydrochloride is an intermediate in the synthesis of Tryptophan derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tryptophan Methyl Ester Hydrochloride is an intermediate in the synthesis of Tryptophan derivatives.

Solubility
Soluble in water (150 mg/ml), dimethyl sulfoxide and methanol (50 mg/ml).

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14907-27-8
Molecular Formula C12H15ClN2O2
Molecular Weight (g/mol) 254.71
MDL Number MFCD00038992
InChI Key XNFNGGQRDXFYMM-UHFFFAOYNA-N
Synonym d-tryptophan methyl ester hydrochloride, h-d-trp-ome.hcl, r-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, methyl 2r-2-amino-3-1h-indol-3-yl propanoate hydrochloride, methyl d-tryptophanate hydrochloride, d-tryptophan methyl ester hcl, h-d-trp-ome hcl, dtrp-ome.hcl, pubchem10879, ksc174m8r
PubChem CID 11448200
IUPAC Name methyl (2R)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
SMILES Cl.COC(=O)C(N)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 213°C to 216°C
Solubility Information Soluble in water (150mg/ml),dimethyl sulfoxide and methanol (50mg/ml).
Quantity 5 g
Formula Weight 254.71
Optical Rotation −18° (c=5 in Methanol)
Percent Purity 98%
Chemical Name or Material D-Tryptophan methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.