missing translation for 'onlineSavingsMsg'
Learn More

D-Tryptophan, 99%, MP Biomedicals™

Product Code. 11453571 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453571 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453571 Supplier MP Biomedicals Supplier No. 0210314401
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity amino acid suitable for research or further manufacturing applications.

Specifications

CAS 153-94-6
Color White
Assay Percent Range 0.99
Molecular Formula C11H12N2O2
MDL Number MFCD00005647
Synonym d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh
InChI Key QIVBCDIJIAJPQS-SECBINFHSA-N
SMILES N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O
IUPAC Name (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 204.23
Quantity 1 g
PubChem CID 9060
ChEBI CHEBI:16296
Formula Weight 204.23
Percent Purity ≥99%
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material D-2-Amino-3(3-indolyl)propionic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.