missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Trehalose dihydrate, MedChemExpress

Product Code. 16489146
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489146 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16489146 Supplier Medchem Express Supplier No. HYN1132A
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-(+)-Trehalose dihydrate, isolated from Saccharomyces cerevisiae, can be used as a food ingredient and pharmaceutical excipient.

D-(+)-Trehalose dihydrate, isolated from Saccharomyces cerevisiae, can be used as a food ingredient and pharmaceutical excipient.

Specifications

Chemical Name or Material D-(+)-Trehalose dihydrate
CAS 6138-23-4
Color White
Purity Grade Notes Research
Molecular Formula C12H26O13
Quantity 100 mg
Synonym D-Trehalose dihydrate α,α-Trehalose dihydrate
Solubility Information H2O : 150 mg/mL (396.48 mM; Need ultrasonic)
SMILES O[C@H]1[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)O[C@H](CO)[C@@H](O)[C@@H]1O.[2 H2O]
Molecular Weight (g/mol) 378.33
Formula Weight 378.33
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.