missing translation for 'onlineSavingsMsg'
Learn More

D-Threonine, TRC

Product Code. 30454966
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454966 5 g 5g
30454753 25 g 25g
30453346 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454966 Supplier Toronto Research Chemicals Supplier No. TRCT4055255G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-20-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
Synonym (2R,3S)-2-Amino-3-hydroxybutanoic Acid, (2R,3S)-3-Hydroxy-2-aminobutanoic Acid, (R)-Threonine, NSC 46702
IUPAC Name (2R,3S)-2-amino-3-hydroxybutanoic acid
SMILES [C@@H]([C@H](C)O)(C(O)=O)N

Specifications

InChI Formula InChI=1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m0/s1
Quantity 5 g
Formula Weight 119.0582
Shipping Condition Room Temperature
Chemical Name or Material D-Threonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.