missing translation for 'onlineSavingsMsg'
Learn More

D-tert-Leucine Methyl Ester Hydrochloride, TRC

Product Code. 30360040
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
Unit Size:
1g
2g
500mg
Product Code. Quantity unitSize
30360040 2 g 2g
30349641 500 mg 500mg
30353624 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 30360040

Brand: Toronto Research Chemicals TRCD2469302G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167223-43-0
Molecular Formula C7H15NO2 . HCl
Molecular Weight (g/mol) 181.66
IUPAC Name methyl (2R)-2-amino-3,3-dimethylbutanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)C(C)(C)C

Specifications

InChI Formula InChI=1S/C7H15NO2.ClH/c1-7(2,3)5(8)6(9)10-4;/h5H,8H2,1-4H3;1H/t5-;/m0./s1
Quantity 2 g
Formula Weight 181.087
Shipping Condition Room Temperature
Chemical Name or Material D-tert-Leucine Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.