missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Tagatose, 99+%

Product Code. 10390944
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

D(-)-Tagatose, >-99.0%, C6H12O6, CAS Number-87-81-0, tagatose, d, d--tagatose, d-lyxo-hex-2-ulose, 3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, unii-t7a20y888y, naturlose, tagatose nf, lyxo-2-hexulose, keto-d-tagatose, 1g, (5% soln. in H2O) clear colorless to almost colorless, 99+%, 01, 4414

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 87-81-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00134449
InChI Key BJHIKXHVCXFQLS-PQLUHFTBSA-N
Synonym 3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, lyxo-2-hexulose, tagatose, d, d--tagatose, keto-d-tagatose, tagatose nf, unii-t7a20y888y, d-lyxo-hex-2-ulose, naturlose
PubChem CID 92092
ChEBI CHEBI:47693
IUPAC Name (3S,4S,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Spezifikation

Color White
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Beilstein 01, 4414
Merck Index 15, 9161
Specific Rotation Condition - 6.00 (20.00°C c=5,water)
Specific Rotation -5° to -7° (20°C, 589nm) (c=2, H2O)
Formula Weight 180.16
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Tagatose
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'