missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Sucrose, 99+%, for analysis

Product Code. p-3737411 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10388943

Brand: Thermo Scientific Chemicals 220902500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose. | CAS: 57-50-1 | C12H22O11 | 342.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• D(+)-Sucrose is a disaccharide carbohydrate
• It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose


Applications

• D(+)-Sucrose can be used for protein and nucleic acid purification
• Its fatty acid esters have laboratory research applications in transdermal systems
• It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Color White
Loss on Drying 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Quantity 250 g
Specific Rotation +66.3° to +67.0° (24°C, 589nm) (c=26, H20)
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Formula Weight 342.29
Percent Purity ≥99.0%
Grade Analysis
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D(+)-Sucrose
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.