missing translation for 'onlineSavingsMsg'
Learn More

D-Sorbitol, TRC

Product Code. 30009669
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009669 50 g 50g
30009561 100 g 100g
17919754 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009669 Supplier Toronto Research Chemicals Supplier No. TRCS67710050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6 H14 O6
Molecular Weight (g/mol) 182.17
Synonym Sorbitol, d-Glucitol, d-sorbitol
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1
Formula Weight 182.079
Percent Purity >95%
Chemical Name or Material D-Sorbitol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.