missing translation for 'onlineSavingsMsg'
Learn More

D-Sorbitol, MedChemExpress

Product Code. p-200101004
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16448286

Brand: Medchem Express HYB0400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-Sorbitol (Sorbitol) is a six-carbon sugar alcohol and can used as a sugar substitute. D-Sorbitol can be used as a stabilizing excipient and/or isotonicity agent, sweetener, humectant, thickener and dietary supplement.

D-Sorbitol (Sorbitol) is a six-carbon sugar alcohol and can used as a sugar substitute. D-Sorbitol can be used as a stabilizing excipient and/or isotonicity agent, sweetener, humectant, thickener and dietary supplement.

Specifications

Chemical Name or Material D-Sorbitol
Physical Form Solid
CAS 50-70-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H14O6
Synonym Sorbitol D-Glucitol
Solubility Information H2O : 100 mg/mL (548.94 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (548.94 mM; Need ultrasonic)
SMILES OC[C@@H]([C@H]([C@@H]([C@@H](CO)O)O)O)O
Molecular Weight (g/mol) 182.17
Formula Weight 182.17
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.