missing translation for 'onlineSavingsMsg'
Learn More

D-Sorbitol, Anhydrous, Cell Culture Reagent, ≥98%, MP Biomedicals™

Product Code. 11432031 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432031 1 kg 1kg
11442031 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432031 Supplier MP Biomedicals Supplier No. 0219474201
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

D-Sorbitol, Anhydrous, Cell Culture Reagent, ≥98% is used for inducing osmotic stress.

  • Used for washing spheroplasts and in isoelectric focusing to minimize endoosmotic flow in agarose gels.
  • May be used to induce osmotic stress.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, glucarine, diakarmon, multitol, sorbilande
PubChem CID 5780
ChEBI CHEBI:17924

Specifications

Melting Point 230°F
Density 1.49 g/cm3 at 59°F
pH 7.0
Boiling Point Very High
Flash Point 542°F
Quantity 1 kg
Solubility Information Water Solubility: 2,750,000 mg/L at 30°C
Optical Rotation −4 to 0 ° (c=2, water)
Vapor Pressure 9.9X10-9 mm Hg at 25°C
Percent Purity ≥98%
Grade Cell Culture Grade
Physical Form Powder
Conductivity <20.0 μS/cm
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.