missing translation for 'onlineSavingsMsg'
Learn More

D-Serine methyleester hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10396534 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10396534 25 g Glass Bottle 25g
10386534 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396534 Supplier Thermo Scientific Acros Supplier No. 396790250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5874-57-7 | Molecular Formula: C23H19Br2N3O3S | Molecular Weight: 577.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5874-57-7
Molecular Formula C23H19Br2N3O3S
Molecular Weight (g/mol) 577.29
MDL Number MFCD00066121
InChI Key LYRWWUCOZBTOPL-UHFFFAOYNA-N
Synonym d-serine methyl ester hydrochloride, h-d-ser-ome.hcl, r-methyl 2-amino-3-hydroxypropanoate hydrochloride, d-serine, methyl ester, hydrochloride, d-serine methyl ester hcl, methyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-ser-ome hcl, cycloserine impurity 2, h-d-ser-ome.hci
PubChem CID 11446470
IUPAC Name methyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1

Specifications

Melting Point 152.0°C to 161.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2CH3·HCl
Specific Rotation −2.5° to −4.5° (20°C, 589nm) (C=4, methanol)
Quantity 25 g
Formula Weight 155.58
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material D-Serine methyl ester hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • D-Serine methyl ester hydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.