missing translation for 'onlineSavingsMsg'
Learn More

D-Serine Methyl Ester Hydrochloride, TRC

Product Code. 30138624
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138624 25 g 25g
30134586 2.5 g 2.5g
30008709 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138624 Supplier Toronto Research Chemicals Supplier No. TRCS27101025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5874-57-7
Molecular Formula C4 H9 N O3 . Cl H
Molecular Weight (g/mol) 155.58
Synonym D-Serine, methyl ester, hydrochloride (1:1), D-Serine, methyl ester, hydrochloride (9CI), Serine, methyl ester, hydrochloride, D- (8CI), (2R)-3-Hydroxy-l-methoxy-l-oxopropan-2-aminium chloride, (R)-2-Amino-3-hydroxypropionic acid methyl ester hydrochloride, (R)-Methyl 2-amino-3-hydroxypropanoate hydrochloride, Methyl D-serinate hydrochloride, D-Serine methyl ester hydrochloride, (R)-Serine methyl ester hydrochloride, Methyl (2R)-2-amino-3-hydroxypropanoate hydrochloride
IUPAC Name methyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)CO

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H9NO3.ClH/c1-8-4(7)3(5)2-6;/h3,6H,2,5H2,1H3;1H/t3-;/m1./s1
Formula Weight 155.0349
Chemical Name or Material D-Serine Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.