missing translation for 'onlineSavingsMsg'
Learn More

D-Serine, 98%

Product Code. 10378120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10378120 25 g Glass Bottle 25g
10023150 5 g Glass Bottle 5g
10402885 100 g Glass Bottle 100g
11921731 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10378120 Supplier Thermo Scientific Acros Supplier No. 227070250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 312-84-5 | C3H7NO3 | 105.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312-84-5
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00004269
InChI Key MTCFGRXMJLQNBG-UWTATZPHSA-N
Synonym d-serine, r-2-amino-3-hydroxypropanoic acid, 2r-2-amino-3-hydroxypropanoic acid, serine d-form, r-serine, serine, d, d-+-serine, h-d-ser-oh, d-2-amino-3-hydroxypropionic acid, d-serin
PubChem CID 71077
ChEBI CHEBI:16523
IUPAC Name (2R)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 220.0°C
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2H
Specific Rotation − 14.75
Solubility Information Solubility in water: soluble in water: 346g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene., soluble in mineral acids., insoluble in organic solvents.
Specific Rotation Condition −14.75° (20°C c=10 2 N HCl)
Quantity 25 g
Formula Weight 105.09
Percent Purity 98%
Chemical Name or Material D-Serine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.