missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Ribose-5-phosphate disodium salt hydrate, 85%

Product Code. 15410657
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15410657

Brand: Thermo Scientific Chemicals J66815.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18265-46-8 | C5H9Na2O8P | 274.07 g/mol

Ribose-5-phosphate is an intermediate of the oxidative branch of the Pentose Phosphate Pathway. Ribose-5-phosphate is used in the synthesis of nucleotides and nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ribose-5-phosphate is an intermediate of the oxidative branch of the Pentose Phosphate Pathway. Ribose-5-phosphate is used in the synthesis of nucleotides and nucleic acids.

Solubility
Soluble in water at 50mg/ml

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18265-46-8
Molecular Formula C5H9Na2O8P
Molecular Weight (g/mol) 274.07
MDL Number MFCD00135931
InChI Key RHCIGTFSCDIJGH-CHEKPSGWSA-L
Synonym d-ribofuranose5-phosphatedisodiumsalt
PubChem CID 131875268
IUPAC Name sodium;[(2R,3R,4R)-2,3,4-trihydroxy-5-oxopentyl] dihydrogen phosphate
SMILES [Na+].[Na+].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O

Specifications

Color White to Yellow
Quantity 250 mg
Solubility Information Soluble in water at 50mg/ml
Formula Weight 310.1
Percent Purity 85%
Physical Form Powder
Chemical Name or Material D-Ribose-5-phosphate disodium salt hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.