missing translation for 'onlineSavingsMsg'
Learn More

D-Ribonolactone, TRC

Product Code. 30008567
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30008567 Supplier Toronto Research Chemicals Supplier No. TRCR4160005G
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5336-08-3
Molecular Formula C5 H8 O5
Quantity 5 g
Synonym D-Ribonic acid, γ-lactone, Ribonic acid, γ-lactone, D- (8CI), (+)-Ribonolactone, D-(+)-Ribonic acid lactone, D-(+)-Ribonic acid γ-lactone, D-(+)-Ribonolactone, D-Ribono-1,4-lactone, D-Ribono-γ-lactone, D-Ribonolactone, D-Ribopentono-1,4-lactone, NSC 1031, Ribonic acid 1,4-lactone
InChI Formula InChI=1S/C5H8O5/c6-1-2-3(7)4(8)5(9)10-2/h2-4,6-8H,1H2/t2-,3-,4-/m1/s1
SMILES OC[C@H]1OC(=O)[C@H](O)[C@@H]1O
IUPAC Name (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
Molecular Weight (g/mol) 148.11
Formula Weight 148.0372
Chemical Name or Material D-(+)-Ribonicacid-gamma-lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.