missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Ribonic acid gamma-lactone, 96%

Product Code. 10781501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10781501 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10781501 Supplier Thermo Scientific Acros Supplier No. 227830100
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5336-08-3 | C5H8O5 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5336-08-3
Melting Point 83.0°C to 85.0°C
Ignition Residue 0.2% max
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C5H8O5
MDL Number MFCD00063241
Quantity 10 g
Specific Rotation Condition + 18.00 (20.00°C c=5,H2O)
Specific Rotation + 18.00
Synonym ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone
Solubility Information Solubility in water: soluble
InChI Key CUOKHACJLGPRHD-UHFFFAOYNA-N
SMILES C(C1C(C(C(=O)O1)O)O)O
IUPAC Name (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
Molecular Weight (g/mol) 148.12
PubChem CID 111064
ChEBI CHEBI:74168
Formula Weight 148.12
Percent Purity 96%
Chemical Name or Material D(+)-Ribonic acid γ-lactone, 96%
Show More Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.