missing translation for 'onlineSavingsMsg'
Learn More

D-Ribonic acid-1,4-lactone

Product Code. 15417559
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417559 5 g 5g
15407559 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417559 Supplier Thermo Scientific Alfa Aesar Supplier No. J66266.06
  • ONLINE EXCLUSIVE PRICE
    £189.00
    £170.65 / 5g
    Save £18.35
    9% Off
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5336-08-3 | C5H8O5 | 148.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5336-08-3
Molecular Formula C5H8O5
Molecular Weight (g/mol) 148.11
MDL Number MFCD00063241
InChI Key CUOKHACJLGPRHD-UHFFFAOYNA-N
Synonym ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone
PubChem CID 111064
ChEBI CHEBI:74168
IUPAC Name (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
SMILES C(C1C(C(C(=O)O1)O)O)O

Specifications

Melting Point 85°C to 87°C
Color White to Yellow
Quantity 5 g
Beilstein 82057
Solubility Information Soluble in water at 100mg/ml
Formula Weight 148.11
Physical Form Powder
Chemical Name or Material D-Ribonic acid-1,4-lactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.