missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-D-Ribofuranose 1-acetate 2,3,5-tribenzoate, 99%

Product Code. 10761441 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10761441 10 g Glass bottle 10g
10513864 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10761441 Supplier Thermo Scientific Chemicals Supplier No. 170110100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6974-32-9 | C28H24O9 | 504.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6974-32-9
Molecular Formula C28H24O9
Molecular Weight (g/mol) 504.48
MDL Number MFCD00005357
InChI Key GCZABPLTDYVJMP-CBUXHAPBSA-N
Synonym 1-o-acetyl-2,3,5-tri-o-benzoyl-beta-d-ribofuranose, beta-d-ribofuranose 1-acetate 2,3,5-tribenzoate, 2r,3r,4r,5s-5-acetyloxy-3,4-bis benzoyloxy oxolan-2-yl methyl benzoate, 1-acetyl-2,3,5-tribenzoyl-beta-d-ribofuranose, 1-acetyl-2,3,5-tribenzoy-b-d-ribofuranose, 1-o-acetyl-2,3,5,tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5s-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl methyl benzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxymethyl tetrahydrofuran-3,4-diyl dibenzoate, .beta.-d-ribofuranose, 1-acetate 2,3,5-tribenzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxy methyl tetrahydrofuran-3,4-diyl dibenzoate
PubChem CID 81455
IUPAC Name [(2R,3R,4R,5S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate
SMILES CC(=O)OC1C(C(C(O1)COC(=O)C2=CC=CC=C2)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.0°C to 130.0°C
Color Brown to Green or Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Specific Rotation Condition + 24.40 (20.00°C c=1,pyridine)
Specific Rotation + 24.40
Solubility Information Solubility in water: insoluble
Formula Weight 504.48
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material β-D-Ribofuranose 1-acetate 2, 3, 5-tribenzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.