missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%

Product Code. 10697522 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697522 25 g Glass bottle 25g
10646214 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10697522 Supplier Thermo Scientific Chemicals Supplier No. 170120250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13035-61-5 | C13H18O9 | 318.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13035-61-5
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.27
MDL Number MFCD00005358
InChI Key IHNHAHWGVLXCCI-FDYHWXHSSA-N
Synonym beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
PubChem CID 83064
IUPAC Name [(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 79°C to 81°C
Color White
Ignition Residue 0.1% max.
Loss on Drying 1% max. (60°C, 2 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 25 g
Specific Rotation Condition −15.40° (20°C c=7,MeOH)
Specific Rotation -15.4
Formula Weight 318.27
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material β-D-Ribofuranose 1,2,3,5-tetraacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.