missing translation for 'onlineSavingsMsg'
Learn More

D-ribo Phytosphingosine, TRC

Product Code. 30179500
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179500 100 mg 100mg
30178924 5 mg 5mg
30179132 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179500 Supplier Toronto Research Chemicals Supplier No. TRCP398990100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-62-1
Molecular Formula C18 H39 N O3
Molecular Weight (g/mol) 317.51
Synonym (2S,3S,4R)-2-Amino-1,3,4-octadecanetriol, Phytosphingosine, (+)-D-ribo-Phytosphingosine, 4-D-Hydroxysphinganine, 4D-Hydroxysphinganine, C18-Phytosphingosine, D-ribo-1,3,4-Trihydroxy-2-aminooctadecane;
IUPAC Name (2S,3S,4R)-2-aminooctadecane-1,3,4-triol
SMILES CCCCCCCCCCCCCC[C@@H](O)[C@@H](O)[C@@H](N)CO

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H39NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-17(21)18(22)16(19)15-20/h16-18,20-22H,2-15,19H2,1H3/t16-,17+,18-/m0/s1
Formula Weight 317.293
Chemical Name or Material D-ribo Phytosphingosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.