missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Raffinose pentahydrate, 98%, ≥98%, MP Biomedicals™

Product Code. 11496172 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496172 500 g 500g
11476172 25 g 25g
11486172 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496172 Supplier MP Biomedicals Supplier No. 0210279790
  • ONLINE EXCLUSIVE PRICE
    £912.00
    £854.70 / 500g
    Save £57.30
    6% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17629-30-0
Molecular Formula C18H42O21
Molecular Weight (g/mol) 594.513
InChI Key BITMAWRCWSHCRW-ORBWWOJFSA-N
Synonym d-+-raffinosepentahydrate
PubChem CID 134129414
IUPAC Name (3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O

Specifications

Color White
Density 1.465g/mL
Assay Percent Range 0.98
Quantity 500 g
Formula Weight 594.5
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Raffinose pentahydrate, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.