missing translation for 'onlineSavingsMsg'
Learn More

D-Proline, TRC

Product Code. 30427186
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427186 100 mg
30423917 1 g
30425926 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427186 Supplier Toronto Research Chemicals Supplier No. TRCP755990100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-25-2
Molecular Formula C5 H9 N O2
Molecular Weight (g/mol) 115.13
Synonym D-Proline, Proline, D- (8CI), (+)-(R)-Proline, (R)-(+)-Proline, (R)-2-Carboxypyrrolidine, (R)-Proline, (R)-Pyrrolidine-2-carboxylic acid, (R)-Pyrrolidine-2-carboxylic acid, D-(+)-Proline
IUPAC Name (2R)-pyrrolidine-2-carboxylic acid
SMILES OC(=O)[C@H]1CCCN1

Specifications

InChI Formula InChI=1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m1/s1
Quantity 100 mg
Formula Weight 115.0633
Shipping Condition Room Temperature
Chemical Name or Material D-Proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.