missing translation for 'onlineSavingsMsg'
Learn More

D-Proline methyl ester hydrochloride, 98%, Thermo Scientific™

Product Code. 11662179 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11662179 5 g 5g
11915411 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11662179 Supplier Thermo Scientific Acros Supplier No. 445590050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 65365-28-8
Molecular Formula C6H11NO2·ClH
Molecular Weight (g/mol) 165.62
InChI Key HQEIPVHJHZTMDP-NUBCRITNSA-N
Synonym d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride
PubChem CID 44629857
IUPAC Name methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
SMILES COC(=O)C1CCCN1.Cl

Specifications

Melting Point 42°C to 49°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5 to max. 102.5% min. (Argentometry)
Packaging Glass Bottle
Quantity 5 g
Specific Rotation 35
Formula Weight 165.62
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material D-Proline methyl ester hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • D-Proline methyl ester hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.