missing translation for 'onlineSavingsMsg'
Learn More

D-Proline Methyl Ester Hydrochloride, TRC

Product Code. 30414274
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414274 1 g
30418248 50 g
30427185 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30414274 Supplier Toronto Research Chemicals Supplier No. TRCP7559501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65365-28-8
Molecular Formula C6H12ClNO2
Molecular Weight (g/mol) 165.62
Synonym D-Proline Methyl Ester Monohydrochloride, Methyl D-Prolinate Hydrochloride, Methyl Prolinate Hydrochloride, Methyl Proline Hydrochloride
IUPAC Name methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
SMILES Cl.COC(=O)[C@H]1CCCN1

Specifications

InChI Formula InChI=1S/C6H11NO2.ClH/c1-9-6(8)5-3-2-4-7-5;/h5,7H,2-4H2,1H3;1H/t5-;/m1./s1
Quantity 1 g
Formula Weight 165.0557
Shipping Condition Room Temperature
Chemical Name or Material D-Proline Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.