missing translation for 'onlineSavingsMsg'
Learn More

D-Proline, ≥99%, MP Biomedicals™

Product Code. 11415392 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415392 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11415392 Supplier MP Biomedicals Supplier No. 0219545605
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 344-25-2
Color White
Assay Percent Range ≥99%
Molecular Formula C5H9NO2
MDL Number MFCD00064317
Quantity 5 g
Synonym d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-proline
InChI Key ONIBWKKTOPOVIA-SCSAIBSYSA-N
SMILES C1CC(NC1)C(=O)O
IUPAC Name (2R)-pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 115.132
PubChem CID 8988
ChEBI CHEBI:16313
Formula Weight 115.1
Physical Form Powder
Chemical Name or Material D-Proline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.