missing translation for 'onlineSavingsMsg'
Learn More

D-Pipecolinic acid, MedChemExpress

Product Code. 16493459
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493459 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16493459 Supplier Medchem Express Supplier No. HYY0181
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-Pipecolinic acid is a normal human metabolite found in human biofluids.

D-Pipecolinic acid is a normal human metabolite found in human biofluids.

Specifications

Chemical Name or Material D-Pipecolinic acid
CAS 1723-00-8
Color White
Purity Grade Notes Research
Molecular Formula C6H11NO2
Quantity 1 g
Solubility Information H2O : 66.67 mg/mL (516.18 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C([C@@H]1NCCCC1)O
Molecular Weight (g/mol) 129.16
Formula Weight 129.16
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.