missing translation for 'onlineSavingsMsg'
Learn More

D-Pipecolinic acid, 97%

Product Code. 15409858
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409858 5 g 5g
15419858 25 g 25g
15429858 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15409858 Supplier Thermo Scientific Alfa Aesar Supplier No. H63255.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1723-00-8 | C6H11NO2 | 129.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1723-00-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00064346
InChI Key HXEACLLIILLPRG-RXMQYKEDSA-N
Synonym r-piperidine-2-carboxylic acid, d-pipecolic acid, d-pipecolinic acid, d +-pipecolinic acid, d-homoproline, 2r-piperidine-2-carboxylic acid, r-pipecolic acid, +-pipecolic acid, d-piperidine-2-carboxylic acid
PubChem CID 736316
ChEBI CHEBI:41582
IUPAC Name (2R)-piperidine-2-carboxylic acid
SMILES C1CCNC(C1)C(=O)O

Specifications

Melting Point 277°C (decomposition)
Quantity 5 g
Beilstein 81094
Formula Weight 129.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material D-Pipecolinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.