missing translation for 'onlineSavingsMsg'
Learn More

D-Pipecolic Acid, TRC

Product Code. 30419183
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419183 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419183 Supplier Toronto Research Chemicals Supplier No. TRCP479751100MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1723-00-8
Molecular Formula C6 H11 N O2
Molecular Weight (g/mol) 129.16
Synonym 2-Piperidinecarboxylic acid, (2R)-, 2-Piperidinecarboxylic acid, (R)-, Pipecolic acid, D-(+)- (8CI), (2R)-2-Piperidinecarboxylic acid, (+)-Pipecolic acid, (+)-Pipecolinic acid, (2R)-Piperidine-2-carboxylic acid, (R)-(+)-2-Piperidinecarboxylic acid, (R)-(+)-Pipecolic acid, (R)-2-Carboxypiperidine, (R)-2-Piperidinecarboxylic acid, (R)-Pipecolic acid, (R)-Pipecolinic acid, D-(+)-Pipecolic acid, D-Homoproline, D-Pipecolic acid, D-Pipecolinic acid, D-Piperidine-2-carboxylic acid
IUPAC Name (2R)-piperidine-2-carboxylic acid
SMILES OC(=O)[C@H]1CCCCN1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H11NO2/c8-6(9)5-3-1-2-4-7-5/h5,7H,1-4H2,(H,8,9)/t5-/m1/s1
Formula Weight 129.079
Chemical Name or Material D-Pipecolinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.