missing translation for 'onlineSavingsMsg'
Learn More

D-Pinitol, TRC

Product Code. 30350012
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
missing translation for 'unitSize'
25g
This item is not returnable. View return policy

Product Code. 30350012

missing translation for 'mfr': Toronto Research Chemicals TRCD21294025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Pinitol
CAS 10284-63-6
Molecular Formula C7 H14 O6
Quantity 25 g
Synonym 3-O-Methyl-D-chiro-inositol, Matezit, Pinit, Sennit, Sennitol
InChI Formula InChI=1S/C7H14O6/c1-13-7-5(11)3(9)2(8)4(10)6(7)12/h2-12H,1H3/t2-,3-,4-,5-,6+,7+/m0/s1
SMILES CO[C@H]1[C@H](O)[C@@H](O)[C@@H](O)[C@H](O)[C@@H]1O
IUPAC Name (1R,2S,4S,5S)-6-methoxycyclohexane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 194.18
Formula Weight 194.079
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.