missing translation for 'onlineSavingsMsg'
Learn More

D-Pinitol, MedChemExpress

Product Code. 16434867
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16434867

Brand: Medchem Express HYN0655

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress D-pinitol (3-O-Methyl-D-chiro-inositol) is a natural compound presented in several plants, like Pinaceae and Leguminosae plants. D-pinitol exerts hypoglycemic activity and protective effects in the cardiovascular system. D-pinitol has antiviral and larvicidal activities.

D-pinitol (3-O-Methyl-D-chiro-inositol) is a natural compound presented in several plants, like Pinaceae and Leguminosae plants. D-pinitol exerts hypoglycemic activity and protective effects in the cardiovascular system. D-pinitol has antiviral and larvicidal activities.

Specifications

Chemical Name or Material D-Pinitol
Physical Form Solid
CAS 10284-63-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C7H14O6
For Use With (Application) Neuroscience-Neuromodulation
Synonym 3-O-Methyl-D-chiro-inositol
Solubility Information DMSO : 125 mg/mL (643.73 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H]([C@H]([C@@H]([C@H]([C@H]1O)O)O)OC)O
Molecular Weight (g/mol) 194.18
Formula Weight 194.18
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.