missing translation for 'onlineSavingsMsg'
Learn More

D-Pinitol, MedChemExpress

Product Code. 16489116
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489116 100 mg 100mg
16434867 50 mg 50mg
16419058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16489116 Supplier Medchem Express Supplier No. HYN0655

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-pinitol (3-O-Methyl-D-chiro-inositol) is a natural compound presented in several plants, like Pinaceae and Leguminosae plants. D-pinitol exerts hypoglycemic activity and protective effects in the cardiovascular system. D-pinitol has antiviral and larvicidal activities.

D-pinitol (3-O-Methyl-D-chiro-inositol) is a natural compound presented in several plants, like Pinaceae and Leguminosae plants. D-pinitol exerts hypoglycemic activity and protective effects in the cardiovascular system. D-pinitol has antiviral and larvicidal activities.

Specifications

Chemical Name or Material D-Pinitol
Physical Form Solid
CAS 10284-63-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H14O6
For Use With (Application) Neuroscience-Neuromodulation
Synonym 3-O-Methyl-D-chiro-inositol
Solubility Information DMSO : 125 mg/mL (643.73 mM; Need ultrasonic)
SMILES O[C@@H]1[C@@H]([C@H]([C@@H]([C@H]([C@H]1O)O)O)OC)O
Molecular Weight (g/mol) 194.18
Formula Weight 194.18
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.