missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Pinitol, 95%

Product Code. 15413158
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413158 250 mg 250mg
15493148 1 g 1g
15403158 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15413158 Supplier Thermo Scientific Chemicals Supplier No. H56648.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10284-63-6 | C7H14O6 | 194.18 g/mol

It is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and  has shown antidiabetic properties in mice. It is  believed to be a salt stress regulator in a wide range of plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a precursor to biologically active fluorinated isosteres of inositol, which show cell growth inhibitory properties and  has shown antidiabetic properties in mice. It is  believed to be a salt stress regulator in a wide range of plants.

Solubility
Soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10284-63-6
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00216659
InChI Key DSCFFEYYQKSRSV-FEPQRWDDSA-N
Synonym d-pinitol, pinitol, 3-o-methyl-d-chiro-inositol, d-+-pinitol, inzitol, pinit, +-pinitol, methylinositol, sennitol, d-ononitol
PubChem CID 164619
IUPAC Name (1S,2S,4S,5R)-6-methoxycyclohexane-1,2,3,4,5-pentol
SMILES COC1[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]1O

Specifications

Melting Point 178°C to 185°C
Quantity 250 mg
Merck Index 14,10361
Solubility Information Soluble in water.
Optical Rotation +60° (c=1 in Water)
Formula Weight 194.18
Percent Purity 95%
Chemical Name or Material D-Pinitol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.