missing translation for 'onlineSavingsMsg'
Learn More

D-​(+)​-​Phenyllactic acid, MedChemExpress

Product Code. 16497796
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16497796 Supplier Medchem Express Supplier No. HY30219
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-​(+)​-​Phenyllactic acid is an anti-bacterial agent, excreted by Geotrichum candidum, inhibits a range of Gram-positive from humans and foodstuffs and Gram-negative bacteria found in humans.

D-​(+)​-​Phenyllactic acid is an anti-bacterial agent, excreted by Geotrichum candidum, inhibits a range of Gram-positive from humans and foodstuffs and Gram-negative bacteria found in humans.

Specifications

Chemical Name or Material D-​(+)​-​Phenyllactic acid
Physical Form Solid
CAS 7326-19-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H10O3
For Use With (Application) COVID-19-immunoregulation
Synonym D-3-Phenyllactic acid
Solubility Information DMSO : 100 mg/mL (601.79 mM; Need ultrasonic)
SMILES O=C(O)[C@H](O)CC1=CC=CC=C1
Molecular Weight (g/mol) 166.17
Formula Weight 166.17
Percent Purity 99.54%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.