missing translation for 'onlineSavingsMsg'
Learn More

D-Phenylalaninol, TRC

Product Code. 30419680
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30419680 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30419680

Brand: Toronto Research Chemicals TRCP3195155G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Phenylalaninol
CAS 5267-64-1
Molecular Formula C9H13NO
Quantity 5 g
Synonym (betaR)-beta-Amino-benzenepropanol, D-2-Amino-3-phenyl-1-propanol, ((1R)-1-Hydroxymethyl-2-phenylethyl)amine, (2R)-2-Amino-3-phenyl-1-propanol, (R)-(+)-2-Amino-3-phenyl-1-glycinol, (R)-2-Amino-1-hydroxy-3-phenylpropane, (R)-2-Benzylethanolamine
InChI Formula InChI=1S/C9H13NO/c10-9(7-11)6-8-4-2-1-3-5-8/h1-5,9,11H,6-7,10H2/t9-/m1/s1
SMILES N[C@@H](CO)Cc1ccccc1
IUPAC Name (2R)-2-amino-3-phenylpropan-1-ol
Molecular Weight (g/mol) 151.21
Formula Weight 151.0997
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.