missing translation for 'onlineSavingsMsg'
Learn More

D-Phenylalanine, TRC

Product Code. 30134399
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134399 25 g 25g
30134233 100 g 100g
30003517 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134399 Supplier Toronto Research Chemicals Supplier No. TRCP31941025G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 673-06-3
Molecular Formula C9 H11 N O2
Molecular Weight (g/mol) 165.19
Synonym D-Phenylalanine, Alanine, phenyl-, D- (8CI), (R)-(+)-Phenylalanine, (R)-2-Amino-3-phenylpropionic acid, (R)-3-Phenyl-2-aminopropanoic acid, (R)-Phenylalanine, D-(+)-Phenylalanine, D-α-Amino-β-phenylpropionic acid, Endorphenyl, NSC 25005
IUPAC Name (2R)-2-amino-3-phenylpropanoic acid
SMILES N[C@H](Cc1ccccc1)C(=O)O

Specifications

InChI Formula InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m1/s1
Quantity 25 g
Formula Weight 165.079
Shipping Condition Room Temperature
Chemical Name or Material D-Phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.